missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ S41i Incubator Shaker Cell Line Bundle (GB)
The New Brunswick S41i is a CO2 incubator with an integrated shaker. An additional perforated shelf that can be used for static incubation at the same time as shaking. Contamination protection is supported by a high temperature disinfection, the fanless design and the seamless chamber.
Brand: Eppendorf™ S41I230112
Description
- mAb-Discovery Bundle: Cell Culture Versatility Solution, for Hybridoma cultures, antibody expression, and cell line establishment
- Bundle consists of: 1x New Brunswick™ S41i, 170 L, (25 mm/1 in. orbit), 1x universal platform and 2x sticky pads, 230 V, 50 Hz
- Experimental flexibility: Perforated shelf allows for static incubation at the same time as shaking
- Stable, uniform and vibration-free shaking: robust Innova triple eccentric drive
- Saves gas and provides an optimal growing atmosphere: Smart gas control (0.2 - 20% CO2)
- Ensures surface sterility: automatic high temperature disinfection and inner/outer door
- Fast and reliable cleaning: Smooth, seamless chamber
- Increasable capacity: Two devices stackable
- Precise and easy control: Intuitive touch screen
- Humidification included
- Shaking speed 25 - 400 rpm (25 - 250 rpm if stacked)
Specifications
25 mm | |
230 V | |
170 L | |
28.7 in. | |
73 cm | |
33.5 in. | |
87.5 cm | |
Touchscreen | |
Continuous |
25 to 400 rpm (25 to 250 rpm if stacked) | |
4°C to 50°C | |
50 Hz | |
17.5 in. | |
44.4 cm | |
34.4 in. | |
69.1 cm | |
Ethernet |