missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Acura™ Electro 926 Micropipette
Lightweight and ergonomically friendly, this electronic single-channel micropipette comes with a QC certificate, Qualitips™ pipette tip samples and operating instructions. Socorex™ Acura™ Electro 926 Micropipette charges quickly and has a long working autonomy.
£545.00 - £795.00
Specifications
Battery Type | 4.8V NiMH |
---|---|
Certifications/Compliance | EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) |
Model | Acura™ Electro™ 926 |
Number of Channels | 1 |
Product Line | Acura™ Electro™ |
Product Code | Brand | Volume (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Description | Price | Quantity & Availability | ||||
11829213
|
Socorex™
926.0010 |
0.5 to 10 μL (Ultra) | Pipette |
£545.00
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11829223
|
Socorex™
926.0200 |
10 to 200 μL | Pipette |
£550.00
Each |
Estimated Shipment: 13-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11849223
|
Socorex™
926.1000 |
50 to 1000 μL | Pipette |
£590.00
Each |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11839223
|
Socorex™
926.0200E |
10 to 200 μL | Pipette Package |
£795.00
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
11809213
|
Socorex™
926.0002 |
0.1 to 2.0 μL | Pipette |
£550.00
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
11849213
|
Socorex™
926.0010Y |
0.5 to 10 μL | Pipette |
£550.00
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
11869213
|
Socorex™
926.0020 |
1.0 to 20 μL | Pipette |
£550.00
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
11889213
|
Socorex™
926.0050 |
2.5 to 50 μL | Pipette |
£550.00
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Exceptional, user-friendly ergonomics.
- Lightweight.
- Easy, intuitive, self-teaching program.
- Large display, reversible right/left reading.
- Microprocessor-controlled motor.
- Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
- Reduced shaft length and conical end improve drivability and access to microtubes and microplates.
- Includes pipetting cycle counter.
- Optional volumetric modules.
Specifications
4.8V NiMH | |
Acura™ Electro™ 926 | |
Acura™ Electro™ | |
Electronic Micropipette | |
1 Each | |
24 Months |
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) | |
1 | |
Self-Teaching | |
Right/Left LCD | |
<1.5Hours |