missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13192-1-AP-150UL
This item is not returnable.
View return policy
Description
Major acute phase reactant. Apolipoprotein of the HDL complex.Specifications
SAA2 | |
Polyclonal | |
Unconjugated | |
SAA2 | |
SAA, SAA1, SAA2, Serum amyloid A protein, serum amyloid A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20209, 6289 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05367, P0DJI9 | |
SAA2 | |
SAA2 Fusion Protein Ag3869 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |