Learn More
Thermo Scientific™ Sabouraud Dextrose Agar with Chloramphenicol
Cultivate and differentiate fungi in clinical samples and environmental samples with this ready-to-use medium.
£23.70 - £29.55
Specifications
Content And Storage | 2°C to 10°C |
---|---|
Format | 90mm Monoplate |
Product Type | Agar |
Packaging Type | Flow Wrapped - Single Layer |
Quantity | 10/Pk. |
Product Code | Brand | Description | Format | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Format | Quantity | Price | Quantity & Availability | |||
12922618
|
Thermo Scientific™
PO0358A |
Sabouraud Dextrose Agar with Chloramphenicol (Deep Fill) | 90mm Monoplate | 10/Pk. |
£29.55
Pack of 10 |
In Stock
Log in to see stock. |
|||
11933672
|
Thermo Scientific™
PO0161A |
Sabouraud Dextrose Agar with Chloramphenicol | 90mm Monoplate | 10/Pk. |
£23.70
Pack of 10 |
In Stock
Log in to see stock. |
|||
Description
Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, using acidic pH Thermo Scientific™ Sabouraud Dextrose Agar with Chloramphenicol.
Fungi maintain their typical cultural appearance and may thus be readily identified according to the standard macroscopic characteristics. Infections caused by Candida spp. are diverse, ranging from superficial and local to deep-seated and disseminated. Candida albicans and Candida glabrata comprise 70 to 80% of yeast isolates recovered from patients with invasive disease.
This culture media, also available in powder form, has applications in environmental monitoring and sterility testing, enabling corrective measures to be implemented rapidly if unacceptable levels of microbial loading indicate lapses in process control.
Specifications
2°C to 10°C | |
Agar | |
10/Pk. |
90mm Monoplate | |
Flow Wrapped - Single Layer |
Safety and Handling
missing translation for 'shelfLife' : 9 Weeks