missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saccharin, sodium salt hydrate, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless to White |
Infrared Spectrum | Authentic |
Assay Percent Range | 25ppm Toluenesulfonamides (GC) max |
Packaging | Plastic Drum |
Quantity | 5 kg |
Beilstein | 27,17 |
Merck Index | 10,8171 |
Solubility Information | Solubility in water: soluble. Other solubilities: slightly soluble in alcohol, practically insoluble in diethyl ether and, chloroform |
Formula Weight | 205.16 |
Percent Purity | 99+% |
Show More |
Safety and Handling
Product Identifier
- Saccharin, sodium salt hydrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'