missing translation for 'onlineSavingsMsg'
Learn More

Saccharin, sodium salt hydrate, 99+%, Thermo Scientific Chemicals

Product Code. 10553391
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82385-42-0
Molecular Formula C7H4NNaO3S
Molecular Weight (g/mol) 205.16
MDL Number MFCD00149605
InChI Key OAZGZMKGRPRHJX-UHFFFAOYSA-M
Synonym saccharin sodium salt hydrate
PubChem CID 131673955
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one;molecular hydrogen;sodium;hydrate
SMILES [Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12

Specifications

Color Colorless to White
Infrared Spectrum Authentic
Assay Percent Range 25ppm Toluenesulfonamides (GC) max
Packaging Plastic bottle
Quantity 1 kg
Beilstein 27,17
Merck Index 10,8171
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol, practically insoluble in diethyl ether and, chloroform
Formula Weight 205.16
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Saccharin, sodium salt hydrate, 99+%
Show More Show Less
Product Identifier
  • Saccharin, sodium salt hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.