Learn More
Fisherbrand™ Saddle Stool
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Color:
Black
Blue
missing translation for 'unitSize'
Each
Description
- Laboratory saddle stool
- Seat height 540-800 mm.
- Comfortable saddle seat, supports the natural S-shape of the spine.
- 3-zone upholstery foam for optimum pressure distribution.
- 3D seat dynamics to ensure active, dynamic sitting.
- Particularly small aluminium base with a diameter of 540 mm and large castors for high mobility.
- Hygienic Design.
- The stool is designed to collect no dirt and to be easy to clean.
Specifications
Specifications
| Product Type | Saddle Stool |
| Color | Blue |
| Material | Steel |
| Seating Type | Stool |
Meets biosafety level S1 and S2, GMP compliant