missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ Safe Aspiration Station
The Safe Aspiration Station is a stand-alone motorized vacuum system for quick and easy aspiration of biological liquid waste disposal. The station offers adjustable aspiration speed and handles. Ideal for ELISA tests, cell culture, and any other contaminated or hazardous liquid aspiration and disposal.
£2173.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | Blue |
Dispensing Speed | 17 mL/s |
Disposable | Reusable |
Grip Type | Ergonomic |
Description
Safe and Contamination-Free
- Closed, stand-alone design to prevent contamination hazards
- All parts in contact with liquids are fully autoclavable
- Equipped with a level detection system to prevent liquid overflow
- Features an in-line hydrophobic filter that traps aerosols for safe disposal
Adaptable
- Various adapters included to be fitted to the hand operator
- Adjustable aspiration level indicated by a board of LEDs
- Modular design enables quick and accurate aspiration of liquids
Convenient
- Easy to use on a bench or in laminar flow cabinets
- Always ready to use by simply pressing the button on the hand operator
- Ergonomic hand operator can be stored on the bottle handle
Specifications
Not Autoclavable | |
17 mL/s | |
Ergonomic | |
1 | |
Non-sterile | |
4 L |
Blue | |
Reusable | |
Polypropylene | |
MLH Product Line | |
Motorized Vacuum |