missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Safe Lock Micro Test Tube
GREENER_CHOICE

Use these polypropylene micro tubes for a variety of applications. Eppendorf™ Safe Lock Micro Test Tubes feature hinged Eppendorf Safe-Lock lids for outstanding protection against unintentional opening during incubation and storage.

£61.90 - £90.06

Specifications

Material Polypropylene
Temperature Range (Metric) –86 to 100°C
View More Specs

Products 6
Product Code Brand Color Capacity (Metric) Price Quantity & Availability  
Product Code Brand Color Capacity (Metric) Price Quantity & Availability  
10663981
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
0030120221
Blue 2 mL
£90.06
Pack of 1000
In Stock
to see stock.
Edge
Add to basket
 
10768371
GREENER_CHOICE
View Documents
Eppendorf™
0030121147
Green 0.5 mL
£61.90
Pack of 500
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
 
10058770
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
0030120213
Red 2 mL
£86.60
Pack of 1000
N/A
Currently Unavailable: We have no information when this product will be available
View alternative products
10547381
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
0030120205
Yellow 2 mL
£86.60
Pack of 1000
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
10421193
GREENER_CHOICE
View Documents
Eppendorf™
0030121112
Yellow 0.5 mL
£64.38
Pack of 500
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
10490993
GREENER_CHOICE
View Documents
Eppendorf™
0030121139
Blue 0.5 mL
£61.90
Pack of 500
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without mold release agents, softeners or biocides that could have a negative effect on bioassays
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • Large frosted lid and surface on the side for easy labeling
  • Guaranteed functionality from -86 to +100°C
  • Autoclavable when open (121°C, 20 min.)
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur quality for guaranteed purity
  • Available in Eppendorf Protein and DNA LoBind material for maximum recovery of valuable samples
Specifications

Specifications

Polypropylene
–86 to 100°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Safe Lock Micro Test Tube

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.