missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Safe-Lock Tubes 2.0 mL - Microtube

Eppendorf™ Safe-Lock Tubes 2.0 mL - Microtube ensures secure sample preservation. With an innovative Safe-Lock mechanism, these microtubes prevent accidental openings, providing reliability in a 2.0 mL capacity. Ideal for various applications, they combine durability, ease of use, and sample protection.
Brand: Eppendorf™ 0030123620
37.99 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- The Eppendorf Safe-Lock tube with a hinged lid prevents accidental opening during incubation and storage.
- g-Safe™ technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30,000 x g, preventing sample loss from tube breakage and enhancing safety when handling hazardous samples.
- Manufactured without the use of slip agents, plasticizers, and biocides – substances known to leach from plastic consumables into the sample, negatively affecting bio-assay results.
- Polypropylene construction imparts high resistance to chemicals, mechanical stress, and extreme temperatures.
- Precise lid sealing minimizes evaporation rates during prolonged storage.
- The tube features a large frosted lid and side surface for easy labeling.
- Functionality is assured within a temperature range from -86 °C to 100 °C.
- Autoclavable when open (121°C, 20 min.) for effective sterilization.
- Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur™ quality for tested purity.

Specifications
Hinged lid | |
Polypropylene | |
2 mL | |
Polypropylene | |
500 Pk. | |
-122.8°F to +212°F | |
Non-sterile | |
Yes | |
Safe-lock Lid | |
Locking Snap-cap Microtube |
Colorless | |
Autoclavable | |
Clear | |
Incubation and storage | |
500 Tubes (10 Bags x 50 Tubes) | |
-86°C to +100°C | |
Forensic DNA Grade | |
0.07 oz. | |
22,000 x g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ Safe-Lock Tubes 2.0 mL - Microtube > Purity Grade: Forensic DNA Grade (Non-sterile), Color: Clear, Packaging: 500 Tubes (10 Bags x 50 Tubes), Max. RCF: 22,000 x g
Spot an opportunity for improvement?Share a Content Correction