missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ SafeCode Plates

Plates with SafeCode barcoding enable safe and reliable sample identification at the bench. Different types of plates available.

Brand:  Eppendorf™ 0030113527

201.00 GBP valid until 2024-12-31
Use promo code "23292" to get your promotional price.



Additional Details : Weight : 2.25000kg

 View more versions of this product

Product Code. 17835938

  • £212.00 / Pack of 20
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • 3-level coding for safe sample and vessel identification
  • Codes on three sides of plates for flexible reading
  • ECC 200 error correction function enables safe code reading with up to 30% damaged codes
  • Additional, ID-specific information on plates available via Eppendorf™ DataPort for documentation purposes
  • Direct import into eLabInventory and eLabJournal software for convenient handling
  • Import to other database systems possible
  • Manufactured from high-quality polypropylene for highest sample purity
Specifications

Specifications

Includes Barcode
8 x 12
Clear
Polypropylene
96
Non-treated
4 Plates/Bag, 5 Bags/Pk.
SBS Standard
Eppendorf
Deepwell Plate
2000 μL
Sterile
Round Bottom (U-well)
PCR Ready
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers