Learn More
DuPont™ Tyvek 500 Xpert Hooded Coverall
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
Brand: DuPont™ D15553613
633.90 GBP valid until 2024-12-31
Use promo code "22918" to get your promotional price.
Additional Details : Weight : 5.00000kg
Description
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
- Robust yet lightweight
- 3-piece hood for optimal fit to head and face when turning
- Elasticated face, wrists and ankles as well as glued-in waist elastic
- Ample crotch area for freedom of movement
- Large, easy-to-grasp zipper puller
- Very low inward leakage thanks to optimized design
Specifications
4X-Large | |
Zipper | |
(EU) 2016/425, EN 14126, EN 1149-5 |
White | |
Stitched |
The user should make the risk assessment to determine the protection required.