missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety Bottle Carrier

Product Code. p-7229412 Shop All Fisherbrand Products
Click to view available options
Color:
Black
Blue
Green
Red
Size:
1.5 L
4.5 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17880772

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in your lab.

  • Robust Design
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
  • Carriers come in four colors with designated labeling areas for easy identification
  • Made of chemical resistant thermoplastic elastomer - PP blend

Specifications

Includes 1 Safety Bottle Carrier vessel and attachable handle
Product Type Bottle Holder
Description Safety Bottle Carrier, 1.5 L, Green
Array 1 x 1
Autoclavable Not Autoclavable
Color Green
Holds Bottles
Material Polypropylene
Positions 1
Shape Cylindrical
Shelves None
Stackable No
Sterility Non-sterile
For Use With (Application) Transportation of bottled regeants
For Use With (Equipment) 1.5 L bottles
Size 1.5 L
Temperature (Metric) Range -20 to 60°C
Length (Metric) 11.4 cm
Width (Metric) 11.4 cm
Height (Metric) 16.5 cm (25.4 cm Height, Handle Extended)
Diameter (Metric) Opening 11.4 cm
Diameter (Metric) Inner 11.4 cm
Weight (Metric) .25 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.