missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System

Product Code. 15947466 Shop All asecos Products
Click to view available options
Configuration:
2 Drawers
2 Shelves, 1 Perforated Insert (285 mm), 1 Bottom Collecting Sump
3 Drawers
3 Shelves, 1 Perforated Insert (375 mm), 1 x 33 L Bottom Collecting Sump
3 Shelves, 1 Perforated Insert (380 mm), 1 33 L Bottom Collecting Sump
3 Shelves, 1 Perforated Insert, 1 Bottom Collecting Sump, 3 Tray Shelves, 1 Bottom Collecting Sump
3 Tray Shelves, 1 Perforated Insert (365 mm), 1 55.5 L Bottom Collecting Sump
4 x 19 L Drawers
4 x 29 L Drawers
4 x 6 L Drawers
6 Drawers, 3 Tray Shelves, 1 Bottom Collecting Sump
6 x 19 L Drawers
6 x 29 L Drawers
6 x 6 L Drawers
Unit Size:
Each
This item is not returnable. View return policy

For indoor use only

Product Code. 15947466

Brand: asecos™ 3253200133695

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of flammable hazardous substances in working areas.

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • User-friendly and safe (model .WDAS): door open arrest system, convenient and safe removal or storage of containers
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration 4 x 6 L Drawers
Depth (Metric) Exterior 749 mm
Depth (Metric) Interior 657 mm
Dimensions (D x W x H) Interior 657 x 450 x 1742 mm
Dimensions (D x W x H) 749 x 596 x 1968 mm
Door Style Wing, Left-hinged
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 1968 mm
Height (Metric) Interior 1742 mm
Includes Door open arrest system
Model S90.196.060.075.WDAS
No. of Doors 1
Surface Coating Powder-coated plain
Weight (Metric) 344 kg
Width (Metric) Exterior 596 mm
Width (Metric) Interior 450 mm
Color Light Gray
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.