missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System

Safe and approved storage of flammable hazardous substances in working areas.

Brand:  asecos™ 3012200133753

 View more versions of this product

Product Code. 15962846

  • £3455.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

For indoor use only

This item is not returnable. View return policy

Description

Description

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • User-friendly and safe (model .WDAS): door open arrest system, convenient and safe removal or storage of containers
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing  locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
616 mm
524 x 1050 x 1072 mm
Wing
1298 mm
Door open arrest system
2
342 kg
1050 mm
Steel
2 x drawer (sheet steel powder-coated)
524 mm
616 x 1196 x 1298 mm
Safe and approved storage of flammable hazardous substances in working areas
1072 mm
S90.129.120.WDAS
Powder-coated plain
1196 mm
Light Gray
Safety storage cabinet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers