missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System
Shop All asecos Products
Click to view available options
Configuration:
2 Drawers
2 Shelves, 1 Perforated Insert (285 mm), 1 Bottom Collecting Sump
3 Drawers
3 Shelves, 1 Perforated Insert (375 mm), 1 x 33 L Bottom Collecting Sump
3 Shelves, 1 Perforated Insert (380 mm), 1 33 L Bottom Collecting Sump
3 Shelves, 1 Perforated Insert, 1 Bottom Collecting Sump, 3 Tray Shelves, 1 Bottom Collecting Sump
3 Tray Shelves, 1 Perforated Insert (365 mm), 1 55.5 L Bottom Collecting Sump
4 x 19 L Drawers
4 x 29 L Drawers
4 x 6 L Drawers
6 Drawers, 3 Tray Shelves, 1 Bottom Collecting Sump
6 x 19 L Drawers
6 x 29 L Drawers
6 x 6 L Drawers
Unit Size:
Each
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe (model .WDAS): door open arrest system, convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Specifications
| Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
| Configuration | 6 x 6 L Drawers |
| Depth (Metric) Exterior | 749 mm |
| Depth (Metric) Interior | 657 mm |
| Dimensions (D x W x H) Interior | 657 x 450 x 1742 mm |
| Dimensions (D x W x H) | 749 x 596 x 1968 mm |
| Door Style | Wing, Left-hinged |
| For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
| Height (Metric) Exterior | 1968 mm |
| Height (Metric) Interior | 1742 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction