Learn More
Justrite™ Safety Can For Liquid Disposal
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Safety Can For Liquid Disposal provides a better solution for the containment of liquid flammable or corrosive waste.
£494.00
Specifications
Product Type | Safety Can |
---|---|
Description | Safety Can for Liquid Disposal |
For Use With (Application) | Liquid Disposal |
Material | HDPE |
Description
High-density polyethylene (HDPE) disposal cans provide superior chemical resistance and leak protection. Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig (0.2 and 0.35 bar). Stainless steel handle and hardware with latch-catch system hold the cap in the open position for easy filling and pouring. Large-mouth opening features a built-in stainless steel flame arrester to guard against outside ignition. The broad-base design provides stability and resists accidental tip over.Specifications
Safety Can | |
Safety Can for Liquid Disposal | |
Liquid Disposal | |
HDPE |