missing translation for 'onlineSavingsMsg'
Learn More

SCAT Safety Canisters

Código de producto. 17181511
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
2.5 L
5 L
10 L
12 L
20 L
30 L
60 L
Closure Size:
GL45
S51
S55
S60/61
S60/61 and GL45
S70/71
S90
S95
Includes:
2 openings
Floater
Mechanical Level Control
UN-X approval
UN-X approval; with inner fluorination
UN-Y approval
UN-Y approval, with inner fluorination
Este artículo no se puede devolver. View return policy
This item is not returnable. View return policy

Our SCAT Safety Canisters are made of high-quality PE-HD material with high chemical resistance for use in the laboratory for safe disposal of chromatographic solvents or other liquid laboratory waste for the highest level of work safety in the laboratory.

  • Made of robust HDPE, which is very resistant to almost all solvents commonly used in the laboratory
  • Smallest canisters have a volume of only 5 L and are ideally suited for use on tables or smaller storage areas
  • Larger canisters are available with volumes of 10, 12, 20, 30 up to 60 L, which can be deposited under tables or in safety cabinets
  • Thread types S55, S60/61, S65, S70/71, S90 and S95 are commonly used in the laboratory, they are all compatible with the SCAT SafetyWasteCaps range for the safe disposal of volatile hazardous substances
  • Special versions available with floats for level indication or with internal fluorination for double protection of the contents, both for permeation from outside to inside and for diffusion from inside to outside

The wide range of non-electrically conductive canisters includes a variety of designs for individual use in the laboratory for the disposal of solvents in combination with the SCAT SafetyWasteCaps to increase your laboratory safety.

Specifications

Autoclavable Not Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Closure Size S55
Color White (Translucent)
Description Canister 10 L, S55, F-HDPE with UN-approval, dimensions (W x H x D): 190 x 310 x 230 mm
Dimensions (L x W x H) 190 x 230 x 310 mm
For Use With (Application) For All Purposes and Suitable for The S.C.A.T™ SafetySystem
Includes UN-Y approval, with inner fluorination
Material HDPE
Product Type Canister
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'