missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Safety Canisters, HDPE-EC

Product Code. 17917250
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
30 L
60 L
Closure Size:
S50
S60/61
S70/71
S90
Includes:
Canister
Canister with Blue Stripes on Grip, without UN-Y Approval
Canister with Floater
Canister with Green Stripes on Grip, without UN-Y Approval
Canister with Red Stripes on Grip, without UN-Y Approval
Canister with UN-Y Approval
Canister with Viewing Strip, UN-Y Approval
Canister with Yellow Stripes on Grip, without UN-Y Approval
This item is not returnable. View return policy
This item is not returnable. View return policy

SCAT™ electrically conductive safety canisters are made of high quality HDPE material. Due to their grounding capability, flying sparks caused by electrical discharge can be prevented when disposing of flammable or explosive laboratory liquids.

  • Made of very robust and electrically conductive HDPE, which is very resistant to solvents commonly used in laboratories
  • Filling quantities from 10 L for use with HPLC systems up to massive 60 L drums for the collection of large quantities of containers (e.g. professional SymLine™ disposal systems)
  • Thread types S50, S60/61, S70/71 and S90 are commonly used in the laboratory and are all compatible
  • Safety Waste Caps range for the safe disposal of volatile, hazardous substances
  • Special safety features such as integrated visibility strips or floats ensure permanent fill level monitoring
  • Practical designs as space-saving canisters allow safe storage of liquid waste even in small areas or in hazardous goods cabinets

Specifications

Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Certifications/Compliance UN-Y approval
Closure Size S60/61
Color Black
Description Canister, 10 Liter, S60/61, PE-HD electrostatic conductive, UN-Y approval, dimensions (W x H x D): 192 x 311 x 232 mm
Dimensions (L x W x H) 192 x 311 x 232 mm
For Use With (Application) Collection of Flammable Liquid Waste
Includes Canister with UN-Y Approval
Material HDPE
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'