Learn More
SCAT HPLC Safety Caps
GREENER_CHOICE
These safety caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.
Brand:  SCAT 107607
106.50 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10348149
  • £134.00 / Each
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High quality materials ensure maximum safety for lab and staff
  • 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
  • Stable construction for highest safety and easy handling
  • Chemically resistant against aggressive organic solvents
  • Attractive design and clear arrangement of the connections
  • Improved design for easy connection of capillaries in various colors
  • Free air valve with change indicator included with every SafetyCap
Specifications
Specifications
Autoclavable
Ground Neck Bottles 29/32 mm
PTFE
Safety Cap II
HPLC Systems
2 PFA Fittings with 3.2 mm OD, 1 Air Valve
NS29/32
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SCAT HPLC Safety Caps > HPLC Safety Caps; Type: Safety Cap II, Thread Size (Metric): NS29/32; Includes: 2 PFA Fittings with 3.2 mm OD, 1 Air Valve; For Use With (Equipment): Ground Neck Bottles 29/32 mm