Learn More
SCAT HPLC Safety Caps
These safety caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.
Brand: SCAT 107030
88.60 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of the connections
- Improved design for easy connection of capillaries in various colors
- Free air valve with change indicator included with every SafetyCap
Specifications
1 PFA Fitting with 3.2 mm OD, 1 Air Valve | |
HPLC Systems | |
PTFE | |
Autoclavable |
GLS80 | |
Safety Cap I | |
Lab Bottles |