missing translation for 'onlineSavingsMsg'
Learn More

SCAT HPLC Safety Caps, V2.0, with Shut-Off
GREENER_CHOICE

Product Code. 16423789 Shop All SCAT Europe Products
Click to view available options
Includes:
1 Fitting, 1 Air Valve, 1 Shut-off
2 Fittings, 1 Air Valve, 1 Shut-off
2 Fittings, 1 Air Valve, 2 Shut-offs
3 Fittings, 1 Air Valve, 2 Shut-offs
3 Fittings, 1 Air Valve, 3 Shut-offs
4 Fittings, 1 Air Valve, 4 Shut-offs
6 Fittings, 1 Air Valve, 6 Shut-offs
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16423789

Brand: SCAT 307219

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

SafetyCap is designed to safety extract solvents when working with any HPLC/UHPLC system. The SCAT Safety Caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours.

  • Luer Lock Adapter
  • Improved fitting design for easier connection
  • Constructed of medical grade PTFE, PFA fittings, and a PPS screw cap
  • Includes space for individually labeling
  • Autocalvable up to 200°C
  • Stable construction for improved handling

Specifications

For Use With (Equipment) Lab Bottles
Includes 2 Fittings, 1 Air Valve, 1 Shut-off
Material PFA, PTFE, PPS
Size (Metric) 3.2 mm O.D. fitting
Thread Size GL 45
For Use With (Application) HPLC Systems
Type Safety Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.