Learn More
Lenz Laborglasintrumente™ Safety Clip Conical Joint Clips
POM safety clip for conical ground joints
Specifications
Quantity 10
Material POM
Color Blue
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11933361
View Documents
Lenz Laborglasintrumente™
01103824
For ground joint NS 24/29 N/A
N/A
11963361
View Documents
Lenz Laborglasintrumente™
01103845
For ground joint NS 45/40 N/A
N/A
Description
Description
  • Height adjustable by variable gear rods, also for use with sleeves with grip collar
  • Outstanding chemical resistance
  • Operating temperature up to approximately 150 °C
Specifications
Specifications
10
POM
Blue