Learn More
Heidolph™ Safety Cover for Heat-On Block
This cover for Heat-On Blocks provides an extra level of protection against burns when heating round bottom flasks.
Brand: Heidolph™ 505-80082-00
This item is not returnable.
View return policy
Description
- Ensures more protection against burns when using Heat-On Block
- Despite reduced danger when using Heat-On Blocks as compared to conventional heating bath, risk of burns is not zero- this safety cover reduces that
- Cover is made of chemical-resistant, easy-to-clean, PTFE
Specifications
White | |
500 mL Round Bottom Flask | |
Heat Protection | |
PTFE |
Product Content Correction
Product Title
Heidolph™ Safety Cover for Heat-On Block > Safety Cover for 500 mL Heat-On Block; For Use With (Application): Heat Protection; For Use With (Equipment): 500 mL Round Bottom Flask; Material: PTFE
Spot an opportunity for improvement?Share a Content Correction