Learn More
Heidolph™ Safety Cover for Heat-On Block
This cover for Heat-On Blocks provides an extra level of protection against burns when heating round bottom flasks.
Brand:  Heidolph™ 505-80081-00
 View more versions of this product
Product Code. 13400049
  • £535.00 / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ensures more protection against burns when using Heat-On Block
  • Despite reduced danger when using Heat-On Blocks as compared to conventional heating bath, risk of burns is not zero- this safety cover reduces that
  • Cover is made of chemical-resistant, easy-to-clean, PTFE
Specifications
Specifications
White
200 to 300 mL Round Bottom Flask
Heat Protection
PTFE