Learn More
Bollé Safety Faceshield
15.48 GBP valid until 2024-12-31
Use promo code "23287" to get your promotional price.
Use promo code "23287" to get your promotional price.
Click to view available options
Model:
B-Line
Clear
Flip Up
Tilting
Description
- Adjustable
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Faceshield |
Frame Color | Gray, Yellow |
Model | Tilting |
No. per Pack | 1 Ea. |
Material | Polycarbonate |
Frame Material | Polypropylene |