Learn More
3M™ Safety Goggles 71347-00011
343.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Lens Tint: Clear
Description
- Provide protection against liquid splash and dust
- Indirect ventilation design
- Strong polycarbonate lens for impact resistance
Specifications
Lens Tint | Clear |
Product Type | Safety Goggles |
Ventilation Details | Indirect vented |
Lens Material | Polycarbonate |