Learn More
SymLine™ Signalbox and Switch Amplifier Accessories
Accessories for the Built-in Signalbox and the Switch Amplifier.
Brand:  SymLine™ 108051
59.87 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10367589
  • £80.00 / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height
  • When container is changed, the alert can be muted via the new touchpad
  • Pumps and other external devices can be conveniently triggered via the integrated interface
  • According to ATEX Directives, Ex-sensors have to be installed in conjunction with a switch amplifier when used in explosion-prone areas (Ex-zones)
  • Signalboxes and switch amplifier must be placed outside of the Ex-zone
  • Monitor two filling levels at once
  • Touchpad
  • Level control through status LEDs
  • Optical and acoustic alarm signals
  • Compatible with Switch AmplifierATEX: A widely used synonym for the ATEX directives of the European Union. The term ATEX is derived from the French abbreviation for ATmosphere EXplosive. Currently, this directive comprises two EU Directives on explosion prevention, namely the ATEX Equipment Directive 2014/34/EU and the ATEX Workplace Directive 1999/92/EC.
Specifications
Specifications
1.9 m
Accessorizing Signalbox and Switch Amplifier
Power Supply
EU Power Supply (220/230 V)
Relay Contact
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SymLine™ Signalbox and Switch Amplifier Accessories > For Use With (Equipment): Relay Contact; Includes: EU Power Supply (220/230 V); Product Type: Power Supply