Learn More
PSF Progressive Safety™ Safety Overshoes Safety Toe Protector
Fine, cold rolled chromium steel toecap
Brand: PSF Progressive Safety™ SAFETOES-XL
Code : 91
Additional Details : Weight : 1.00000kg
Description
- 200 joule toe protection
- Manufactured to EN345
- Fine, cold rolled chromium steel toecap
- Black
Specifications
X-Large |