missing translation for 'onlineSavingsMsg'
Learn More

KARTELL Safety Shield

Product Code. p-200004187
Click to view available options
Thickness (Metric):
5 mm
9 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10061081

Brand: KARTELL 0057100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Safety Shield is free standing, portable and stables, with non-skid feet to provide facial protection from hazardous materials.

  • The safety shields are available in two sizes with different thicknesses and are made of high quality, clear acrylic; are free standing, portable and stable, and are all fitted with non -skid feet.
  • Art. 571 is designed to provide laboratory and healthcare personnel with facial protection from hazardous substances, aerosol and splashing.

Specifications

Height (English) 14.37 in.
Height (Metric) 365 mm
Thickness (English) 0.2 in.
Thickness (Metric) 5 mm
Width (English) 9.53 in.
Width (Metric) 242 mm
Product Type Countertop Shield
Material Acrylic
Disposable Reusable
Lens Material None
Frame Material No Frame
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.