Learn More
SCAT Safety Spouts
Safety pourer with integrated screw cap for safe dispensing of liquids.
Brand:  SCAT 610500
13.33 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10399309
  • £16.50 / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
  • Suitable for politainer waste container
  • Flexible and rigid versions available
  • For the usual canister threads S55, S60/61 and S40/71
  • Made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation
  • Specifications
    Specifications
    S55
    HDPE
    HDPE
    Rigid
    Canister, Container & Bottles
    Safety Spout