Learn More
SCAT Safety Spouts
Product Code. 10639880 Shop All SCAT Europe Products
Click to view available options
Closure Size:
S55
S60/61
S70/71
Description:
Fixed
Flexible
Rigid
Rigid with Vent
Unit Size:
Each
Product Code. 10639880
Brand: SCAT 610502
Only null left
Add to basket
Add to basket
Safety pourer with integrated screw cap for safe dispensing of liquids.
  • Suitable for politainer waste container
  • Flexible and rigid versions available
  • For the usual canister threads S55, S60/61 and S40/71
  • Made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation
  • Specifications
    Closure Size S60/61
    Description Fixed
    Liner Material HDPE
    For Use With (Equipment) Canister, Container & Bottles
    Material HDPE
    Product Type Safety Spout