missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Cabinet for Acids and Alkalis SL-CLASSIC
Safe and approved storage of aggressive, non flammable hazardous materials in working areas.
£2024.60 - £3557.36
Specifications
Certifications/Compliance | DIN EN 16121/16122 conformity, GS certified |
---|---|
Depth (Metric) Exterior | 603 mm |
Depth (Metric) Interior | 570 mm |
Color | Light Gray |
Material | Chipboard, Polypropylene |
Alert:
For indoor use only
Product Code | Brand | Dimensions (D x W x H) Interior | No. of Shelves | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (D x W x H) Interior | No. of Shelves | Price | Quantity & Availability | ||||
15231035
|
asecos™
3060600130608 |
570 x 485 x 1869 mm | None |
£3557.36
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12643515
|
asecos™
3061200130615 |
570 x 485 x 914 mm | 6 |
£2251.44
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15221035
|
asecos™
3060600130607 |
570 x 485 x 1869 mm | 8 |
£3103.68
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11959636
|
asecos™
3061200130614 |
570 x 485 x 914 mm | 4 |
£2024.60
Each |
Estimated Shipment: 26-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Safe and approved storage of aggressive, non flammable hazardous materials in working areas
Function / construction
- Robust construction and longevity: cabinet body made of powder-coated sheet steel, safety elements assembled outside the storage compartment for increased protection against corrosion
- Two in one: two hermetically separated storage compartments allow the isolated storage of acids and bases in one cabinet
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- ventilation: integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
Certificates
- type tested in accordance with EN 14727
- EN 16121/16122 conformity
- GS certified
Warranty
Five years
Specifications
DIN EN 16121/16122 conformity, GS certified | |
570 mm | |
Chipboard, Polypropylene | |
Safe and approved storage of aggressive, non fla mmable hazardous materials in working areas | |
2 | |
Melamine resin laminated |
603 mm | |
Light Gray | |
Cabinet for acids and alkalis | |
1965 mm | |
Pull-out | |
485 mm |