missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Safety Stripe Tape 5702
Click to view available options
Color:
Black, Yellow
Unit Size:
12 rolls
Beskrivning
- Applies instantly and is ready to use immediately
- Black and yellow stripes for hazardous areas
- Easy to apply, stretches around corners for a reliable, tight fit
- Vivid yellow and black stripes are locked into vinyl for permanent high visibility
- High durability, abrasion and solvent resistance
- Resists scuffing, moisture, and weathering for long service life
- Strong bond to most floors, great holding power
- One-piece, clean removal from many surfaces for lower clean-up costs
- Used to identify and mark traffic areas and physical hazards
Specifikationer
Specifikationer
| Dimensions (L x W) | 33 m x 50 mm |
| For Use With (Application) | Marking Hazardous Areas |
| Packaging Quantity | 12 Rolls |
| Thickness | 0.11 mm |
| Adhesive Type | Rubber-based Adhesive |
| Color | Black, Yellow |
| Type | Safety Tape |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering