Learn More
SCAT™ Safety Waste Cap 'LISA', V3.0 Electronic Level Control Extension
With the extension Level Control, you can use your LISA to monitor the fill level of your disposal container at any time.
£108.00 - £198.00
Specifications
For Use With (Application) | HPLC Systems |
---|---|
For Use With (Equipment) | SCAT™ Safety Waste Cap 'LISA', V3.0 |
Product Code | Brand | Material | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Type | Price | Quantity & Availability | ||||
17611795
|
SCAT™
350121 |
PTFE, Conductive | Electronic Control |
£176.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
17681735
|
SCAT™
450121 |
PTFE | Electronic Control |
£198.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
17883530
|
SCAT™
450122 |
PTFE, Conductive | Level Control |
£132.00
Pack of 2 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
17873530
|
SCAT™
350122 |
PTFE | Level Control |
£108.00
Pack of 3 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
Description
LISA is the first modular system for HPLC waste, providing occupational safety and environmental protection with proven SCAT™ technology and maximum flexibility. Simply add connections and components as required so you can collect solvents, acids and alkalis safely at any time.
- Simply unscrew the blind plug of the LISA, insert the level control, and connect cable to the signal box
- Permanently displays the fill level in your disposal container and alerts when reached as soon as it is full
- Available in electrically conductive and non-electrically conductive versions
Specifications
HPLC Systems | |
SCAT™ Safety Waste Cap 'LISA', V3.0 |