Learn More
SymLine™ Safety Waste Caps
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
Brand:  SymLine™ 106478
260.48 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15960386
  • £334.00 / Each
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available with different thread sizes for attachment to pipe system
  • Different numbers of hose connections for disposal of liquid waste
  • Available with electronic level sensors that indicate the degree of filling of the disposal container
  • By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Specifications
Waste Disposal
Electronic Level Control, GL25 (m), Immersed Filling Lance, Connector for Exhaust Filter
Safety Waste Cap
SymLine System
S55