Learn More
SymLine™ Safety Waste Caps
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
Brand: SymLine™ 306491
59.06 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available with different thread sizes for attachment to pipe system
- Different numbers of hose connections for disposal of liquid waste
- Available with electronic level sensors that indicate the degree of filling of the disposal container
- By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Waste Disposal | |
3 x PFA-fittings for Capillaries with (OD) 1.6/2.3/3.2 mm, Collector for HPLC Waste | |
Safety Waste Cap |
SymLine System | |
GL45 |