missing translation for 'onlineSavingsMsg'
Learn More

SCAT HPLC Safety Waste Caps, V2.0
GREENER_CHOICE

Product Code. 16413809 Shop All SCAT Europe Products
Click to view available options
Size:
1 x NPT 3/8 in. (f)
2 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID
2 x PFA-fitting 2.3/3.2 mm OD
3 x 1.6/2.3/3.2 mm OD
3 x 2.3/3.2 mm OD
3 x PFA-fitting 2.3/3.2 mm OD
4 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID
4 x PFA-fitting 1.6/2.3/3.2 mm OD
Thread Size:
GL 45
S40/GL40
S60
S60/61
Includes:
1 PFA blind plug, connector for exhaust filter
2 PFA blind plug, connector for exhaust filter
3 PFA and 3 PTFE blind plugs, connector for exhaust filter
4 PFA and 1 PTFE blind plugs, connector for exhaust filter
Connector for exhaust filter
Connector for exhaust filter (45° beveled)
Grounding connection, connector for exhaust filter
Unit Size:
1 sheet
Each
This item is not returnable. View return policy

Product Code. 16413809

Brand: SCAT 307912

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

SCAT Safety Waste Caps protect staff and the environment from harmful solvent vapours. With a suitable exhaust filter, they turn your disposal container into a closed system without escaping solvent vapours.

  • Discover the flexibility of the world's leading system for solvent safety
  • SafetyWasteCap with S 60/61 thread in the new design of generation 2.0
  • Optimum protection when collecting hazardous or toxic liquid waste
  • Flexible system with fittings and tube connectors in different sizes
  • With multiple options to connect different tubing and capillary sizes
  • Time saving in changing the waste containers while tubes are fixed
  • Resistant against aggressive media
  • PTFE Body 360° freely rotatable: Provides easy container change without distortion of tubing
  • Body with integrated FKM Seal: Provides hermetical connection to the container opening and adapts smoothly even to rugged glass surfaces. An additional PTFE layer protects the seal from direct contact to solvents.
  • PFA Fittings: Improved design for easy tubing connection - Excellent chemical resistance - Flammability classification V-0 (UL-94)
  • Medical Grade PTFE: Excellent chemical resistance Food safe FDA compliant -
  • Flammability classification V-0 (UL-94)
  • Insertion Label: Mark your bottles with individual descriptions
  • PPS Screw Cap: Autoclavable / sterilisable up to 200°C - Flammability classification V-0 (UL-94)
  • Ergonomic Design: Enhanced stability and construction - Improved handling - Easy installation - Smart exchange
  • Huge range of accessories, optional: exhaust filters, funnel use, level control preventing overflowing containers

Specifications

For Use With (Equipment) Waste Disposal Container
Includes 1 PFA blind plug, connector for exhaust filter
Material PFA, PTFE, PPS
Size 3 x PFA-fitting 2.3/3.2 mm OD
Thread Size GL 45
For Use With (Application) Waste Disposal System
Type Safety Waste Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.