missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT HPLC Safety Waste Caps, V2.0
Shop All SCAT Europe Products

Click to view available options
Size:
1 x NPT 3/8 in. (f)
2 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID
2 x PFA-fitting 2.3/3.2 mm OD
3 x 1.6/2.3/3.2 mm OD
3 x 2.3/3.2 mm OD
3 x PFA-fitting 2.3/3.2 mm OD
4 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID
4 x PFA-fitting 1.6/2.3/3.2 mm OD
Thread Size:
GL 45
S40/GL40
S60
S60/61
Includes:
1 PFA blind plug, connector for exhaust filter
2 PFA blind plug, connector for exhaust filter
3 PFA and 3 PTFE blind plugs, connector for exhaust filter
4 PFA and 1 PTFE blind plugs, connector for exhaust filter
Connector for exhaust filter
Connector for exhaust filter (45° beveled)
Grounding connection, connector for exhaust filter
Unit Size:
1 sheet
Each
Description
- Discover the flexibility of the world's leading system for solvent safety
- SafetyWasteCap with S 60/61 thread in the new design of generation 2.0
- Optimum protection when collecting hazardous or toxic liquid waste
- Flexible system with fittings and tube connectors in different sizes
- With multiple options to connect different tubing and capillary sizes
- Time saving in changing the waste containers while tubes are fixed
- Resistant against aggressive media
- PTFE Body 360° freely rotatable: Provides easy container change without distortion of tubing
- Body with integrated FKM Seal: Provides hermetical connection to the container opening and adapts smoothly even to rugged glass surfaces. An additional PTFE layer protects the seal from direct contact to solvents.
- PFA Fittings: Improved design for easy tubing connection - Excellent chemical resistance - Flammability classification V-0 (UL-94)
- Medical Grade PTFE: Excellent chemical resistance Food safe FDA compliant -
- Flammability classification V-0 (UL-94)
- Insertion Label: Mark your bottles with individual descriptions
- PPS Screw Cap: Autoclavable / sterilisable up to 200°C - Flammability classification V-0 (UL-94)
- Ergonomic Design: Enhanced stability and construction - Improved handling - Easy installation - Smart exchange
- Huge range of accessories, optional: exhaust filters, funnel use, level control preventing overflowing containers
Specifications
Specifications
| Diameter (Metric) Inner | 55.5 mm |
| Length (Metric) | 61 mm |
| For Use With (Equipment) | Waste Disposal Container |
| Includes | Connector for exhaust filter |
| Material | PTFE |
| Size | 4 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID |
| Thread Size | S60/61 |
| Thread Size (Metric) | S60/61 |
| For Use With (Application) | Waste Disposal System |
| Type | Safety Waste Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction