Learn More
SCAT HPLC Safety Waste Caps, V2.0
SCAT Safety Waste Caps protect staff and the environment from harmful solvent vapours. With a suitable exhaust filter, they turn your disposal container into a closed system without escaping solvent vapours.
Brand: SCAT 307108
80.45 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.31100kg
Description
- Discover the flexibility of the world's leading system for solvent safety
- SafetyWasteCap with S 60/61 thread in the new design of generation 2.0
- Optimum protection when collecting hazardous or toxic liquid waste
- Flexible system with fittings and tube connectors in different sizes
- With multiple options to connect different tubing and capillary sizes
- Time saving in changing the waste containers while tubes are fixed
- Resistant against aggressive media
- PTFE Body 360° freely rotatable: Provides easy container change without distortion of tubing
- Body with integrated FKM Seal: Provides hermetical connection to the container opening and adapts smoothly even to rugged glass surfaces. An additional PTFE layer protects the seal from direct contact to solvents.
- PFA Fittings: Improved design for easy tubing connection - Excellent chemical resistance - Flammability classification V-0 (UL-94)
- Medical Grade PTFE: Excellent chemical resistance Food safe FDA compliant -
- Flammability classification V-0 (UL-94)
- Insertion Label: Mark your bottles with individual descriptions
- PPS Screw Cap: Autoclavable / sterilisable up to 200°C - Flammability classification V-0 (UL-94)
- Ergonomic Design: Enhanced stability and construction - Improved handling - Easy installation - Smart exchange
- Huge range of accessories, optional: exhaust filters, funnel use, level control preventing overflowing containers
Specifications
3 x 2.3/3.2 mm OD + connector for exhaust filter | |
Waste Disposal System | |
Waste Disposal Container |
S40/GL40 | |
Safety Waste Cap | |
S40/GL40 |