missing translation for 'onlineSavingsMsg'
Learn More

SCAT HPLC Safety Waste Caps, V2.0

SCAT Safety Waste Caps protect staff and the environment from harmful solvent vapours. With a suitable exhaust filter, they turn your disposal container into a closed system without escaping solvent vapours.

Brand:  SCAT 307916

142.29 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.



Additional Details : Weight : 0.44600kg

 View more versions of this product

Product Code. 17175501

  • £168.00 / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

  • Discover the flexibility of the world's leading system for solvent safety
  • SafetyWasteCap with S 60/61 thread in the new design of generation 2.0
  • Optimum protection when collecting hazardous or toxic liquid waste
  • Flexible system with fittings and tube connectors in different sizes
  • With multiple options to connect different tubing and capillary sizes
  • Time saving in changing the waste containers while tubes are fixed
  • Resistant against aggressive media
  • PTFE Body 360° freely rotatable: Provides easy container change without distortion of tubing
  • Body with integrated FKM Seal: Provides hermetical connection to the container opening and adapts smoothly even to rugged glass surfaces. An additional PTFE layer protects the seal from direct contact to solvents.
  • PFA Fittings: Improved design for easy tubing connection - Excellent chemical resistance - Flammability classification V-0 (UL-94)
  • Medical Grade PTFE: Excellent chemical resistance Food safe FDA compliant -
  • Flammability classification V-0 (UL-94)
  • Insertion Label: Mark your bottles with individual descriptions
  • PPS Screw Cap: Autoclavable / sterilisable up to 200°C - Flammability classification V-0 (UL-94)
  • Ergonomic Design: Enhanced stability and construction - Improved handling - Easy installation - Smart exchange
  • Huge range of accessories, optional: exhaust filters, funnel use, level control preventing overflowing containers
Specifications

Specifications

3 x 2.3/3.2 mm OD, grounding connection + connector for exhaust filter
Safety Waste Cap
S60/61
Waste Disposal System
Waste Disposal Container
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers