Learn More
SCAT™ HPLC Safety Caps, V2.0
These HPLC Safety Caps allow you to extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.
£93.80 - £147.00
Specifications
For Use With (Application) | HPLC Systems |
---|---|
For Use With (Equipment) | Lab Bottle |
Product Code | Brand | Includes | Material | Thread Size | Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Material | Thread Size | Type | Price | Quantity & Availability | ||
17172036
|
SCAT™
307100 |
1 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve | PTFE | GL/S40 | Safety Cap I |
£103.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
17843530
|
SCAT™
307109 |
1 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve | PTFE | GL/S40 | Safety Waste Cap I |
£99.30
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16403789
|
SCAT™
307019 |
1 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve | PTFE | GL 45 | Safety Cap I |
£117.00
Each |
In Stock
Log in to see stock. |
||
17833530
|
SCAT™
307019-PP |
1 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve | Polypropylene | GL 45 | Safety Cap I |
£93.80
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
16453819
|
SCAT™
399019 |
1 x PFA 1.6/2.3/3.2 mm OD Fittings, 90° Angled Air Valve | PTFE | GL 45 | Safety Cap I |
£132.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
17112576
|
SCAT™
307101 |
2 x PFA 1.6/2.3/3.2 mm OD Fittings, PFA Blind Plug, Air Valve | PTFE | GL/S40 | Safety Cap II |
£111.00
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17150711
|
SCAT™
307006 |
2 x PFA 1.6/2.3/3.2 mm OD Fittings, PFA Blind Plug, Air Valve | PTFE | GL 28 | Safety Cap II |
£111.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
16403809
|
SCAT™
307909 |
2 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve | PTFE | GL 45 | Safety Cap II |
£131.00
Each |
In Stock
Log in to see stock. |
||
16483819
|
SCAT™
399909 |
2 x PFA 1.6/2.3/3.2 mm OD Fittings, 90° Angled Air Valve | PTFE | GL 45 | Safety Cap II |
£143.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||
16439423
|
SCAT™
307910 |
3 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve | PTFE | GL 45 | Safety Cap III |
£147.00
Each |
In Stock
Log in to see stock. |
||
16463799
|
SCAT™
307410 |
4 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve | PTFE | GL 45 | Safety Cap IV |
£143.45
Each |
In Stock
Log in to see stock. |
||
17848733
|
SCAT™
307410-PP |
Air Valve, Change Indicator, 4 PFA Fittings (1.6 mm OD), 4 PFA Fittings (2.3 mm OD), 4 PFA Fittings (3.2 mm OD), 3 PFA Blind Plug, Labeling Field | Polypropylene | GL 45 | Safety Cap IV |
£121.00
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||
16493799
|
SCAT™
307520 |
6 x PFA 1.6/2.3/3.2 mm OD Fittings, Air Valve | PTFE | GL 45 | Safety Cap VI |
£132.00
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||
17868733
|
SCAT™
309035 |
1 x PTFE 4 mm OD Fitting, Air Valve | PTFE | GL 45 | Safety Cap I |
£111.00
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||
18686488
|
SCAT™
307909-PP |
Safety Cap II: 1 x air valve, 1 x change indicator, 2 x PP fitting 1.6/2.3/3.3 mm OD, 1 x PP blind plug, 1 x labelling field | Polypropylene | GL45 | Safety Cap |
£103.59
1 sheet |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
18616498
|
SCAT™
307910-PP |
Safety Cap III: 1 x air valve, 1 x change indicator, 3 x PP fitting 1.6/2.3/3.3 mm OD, 2 x PP blind plug, 1 x labelling field | Polypropylene | GL45 | Safety Cap |
£109.05
1 sheet |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
SCAT™ Safety Caps V2.0 provide maximum safety for your HPLC solvent supply.
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of connections
- Improved design for easy connection of capillaries in various colors
- Free air valve with change indicator included with every Safety Cap
Specifications
HPLC Systems | |
Lab Bottle |