missing translation for 'onlineSavingsMsg'
Learn More

SCAT Safety Waste Set V2.0

Safety Waste Set V2.0 includes everything need to get your waste fluid collection site ready for operation quickly.

Brand:  SCAT 307318

84.93 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 16679085

  • £123.00 / 1 set
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Collect liquid waste safely with the SCAT. safety funnel MARCO V2.0
  • Ready to use
  • Consists of lid funnel and different canister sizes in a complete set
Specifications

Specifications

140 mm
HDPE
Set of Canister and Lid Funnel MARCO, V2.0
Safety funnel with lid 'MARCO', V2.0, 10 L Canister
GL45
200 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers