missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Safranin Stain

Use for Gram staining of bacteria. Pro-Lab Diagnostics™ Safranin Stain is designed to be ready to use.

Brand:  Pro-Lab Diagnostics™ PL.7012


 View more versions of this product

Product Code. 12956297

  • £65.65 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions in a selection of convenient and economical sizes.
  • Manufactured in accordance with all current regulatory requirements
  • CE labelled
  • Strict quality control and filtering techniques prior to release
  • Also available are a range of convenient staining kits for Gram, TB and Cryptosporidium staining
Specifications

Specifications

Reagent
500mL
Liquid
Dark Red
Ethanol 10 - <25% , Methanol 1 - <2.5% , Glycerol 1 - <2.5%
Gram Stain
Safranin
(EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark
For Gram staining
78 to 100°C
<20°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Safranin

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, methanol

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pro-Lab Diagnostics™ Safranin Stain > Volume: 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.