missing translation for 'onlineSavingsMsg'
Learn More
Safranine, Pure, C.I.50240, Fisher Chemical Product Code.: 10366050

Safranine, Pure, C.I.50240, Fisher Chemical

Product Code. 10366050
25 g, Glass Jar
Click to view available options
Quantity:
25 g
Packaging:
Glass Jar
This item is not returnable. View return policy

Product Code. 10366050

Brand: Fisher Chemical S/0120/46

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 477-73-6
Color Brown
Packaging Glass Jar
Molecular Formula C20H19ClN4
MDL Number MFCD00011759
Quantity 25 g
Synonym basic red 2, safranine o, gossypimine, safranin, safranine t, safranin o, safranin t, safranine, tolusafranine, hidaco safranine
InChI Key QRYAEWIQIBAZOJ-UHFFFAOYSA-N
SMILES [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
IUPAC Name 3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride
Molecular Weight (g/mol) 350.85
PubChem CID 2723800
Formula Weight 350.6128g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • Safranine O
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Safranine, Pure, C.I.50240, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.