missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SafTest™ Dispenser Set
Dispenser Set is an accessory for SafTest™ test kits.
Brand: MP Biomedicals™
Additional Details : Weight : 3.00000kg
Includes: One dispenser for each reagent (A, B, and preparation Reagent) and one reagent holding rack
Description
- Dispenser set for each chosen endpoint
- Allows for accurate testing, every time
- For use with SafTest™ kits
Specifications
Malonaldehyde Testing | |
Room temperature |