missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SafTest™ Preparation Reagent

Contains replacement SafTest™ Preparation Reagent to run all Saftest™ test kits.

Brand:  MP Biomedicals™ 07PREP2000

Product Code. 15702652

  • £101.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For use with SafTest analyzer
  • Used with all six SafTest test kits
  • Appropriate for all testing endpoints
  • Research area: Lipid degradation and oxidation
  • Detection method: Bichromatic photometer
  • Application area: Oil/fat degradation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Oil/Fat Degradation
Bichromatic Photometer
1 Ea.
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers