missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SafTest™ Test Kit

SafTest™ Test Kit measures percentage analytes in given samples. Choose from 6 different kits for use with SafTest instrument and consumable platforms and detection kits.

Brand:  MP Biomedicals™ 07KTFA2000

 View more versions of this product

Product Code. 15742642

  • £438.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Reagent A, Reagent B, Reagent C, Calibrators
This item is not returnable. View return policy

Description

Description

MP Biomedicals' Test Kits contain all reagents and calibrators necessary to run samples. A control is also included in each Test Kit. Each kit will allow for testing of approximately 100 samples. Sample dilution will likely be required for appropriate analysis.

  • Choose from 6 different kits
  • For use with SafTest instrument and consumable platforms
  • Results in as little as 30 minutes
  • Detection method: Bichromatic Photometer
  • Test Kits contain all reagents and calibrators necessary to run samples.
  • A control is also included in each Test Kit.
  • Each kit will allow for testing of approximately 100 samples.
  • Sample dilution will likely be required for appropriate analysis.
    • Alkenal Test Kit
      • Research Area: Lipid degradation and oxidation
      • Contains all reagents, calibrators
    • Free Fatty Acid Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Calibrators, Control Analyte(s)
      • Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.1 - 2.01% free fatty acid
    • Malonaldehyde Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Calibrators, Control
      • Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.00 to 0.32 mg/kg of sample
    • Percent Fat Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Calibrators, Control Analytes(s)
      • Material Required: SafTest Instrument Platform
      • Quantitation Range: 0.06 to 0
    • Peroxide Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Reagent C, Calibrators, Control Analyte(s)
      • Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.00 to 0.200 MEQ/kg of sample
    • Total Glycerin Test Kit
      • Research Area: Biodiesel
      • Components: Reagent A, Reagent B, Calibrators, Control
      • Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.008 to 0.06% of sample
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Free Fatty Acid Testing
100 Tests
1 Ea.
Bichromatic Photometer
Free Fatty Acid
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.