missing translation for 'onlineSavingsMsg'
Learn More

R&L Enterprises™ Bright Stainless Steel Rods

An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Bright Stainless Steel Rods are durable and corrosion resistant.

Brand:  R&L Enterprises™ 3108/43

 View more versions of this product

Product Code. 11705396

  • £65.15 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Threaded
  • Length of rod does not include the thread
Note:  Includes rod, but not the base.

Specifications

Specifications

Stainless Steel
12.5 mm
Rod
Threaded
1000 mm
Includes rod, but not the base.
10 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.