Learn More
Saint-Gobain Chemfluor™ Fluoropolymer Tubing
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
Brand: Saint-Gobain TSFE14025006250
Code : NEW
Additional Details : Weight : 1.20000kg
Description
- Contains no additives or plasticizers
- Inert to virtually all chemicals
- Nonprotein binding
Order Info
In packs of 50 ft. (15m)
Compliance
Complies with FDA criteria
Specifications
Complies with FDA criteria | |
3.2 mm | |
1/4 in. | |
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals |
0.13 in. | |
Fluoropolymer | |
6.4 mm | |
417 psi |